Learn More
Apexbio Technology LLC 3X (DYKDDDDK) Peptide, 25mg. Cas: MFCD: NA. Synthetic peptide tag
SDP
Supplier:  Apexbio Technology LLC A600125
Encompass_Preferred
3X FLAG Peptide is a synthetic peptide with a 3-time repeated DYKXXD motif. Purity = 98.66%; ≥25mg/ml in TBS (0.5M Tris-HCl, pH 7.4, with 1M NaCl); MW 2861.87
Catalog No. NC1279517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.