Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Piperazinyl)benzamidoxime, 97%, Thermo Scientific™

Catalog No. AAH5836803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5836803 1 g
AAH5836806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5836803 Supplier Thermo Scientific Chemicals Supplier No. H5836803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 749188-33-8
Molecular Formula C11H16N4O
Molecular Weight (g/mol) 220.276
MDL Number MFCD22989358
InChI Key IWRIWCGOFIEEIB-UHFFFAOYSA-N
Synonym 4-1-piperazinyl benzamidoxime
PubChem CID 121235633
IUPAC Name N'-hydroxy-4-piperazin-1-ylbenzenecarboximidamide
SMILES C1CN(CCN1)C2=CC=C(C=C2)C(=NO)N

Specifications

Quantity 1 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 4-(1-Piperazinyl)benzamidoxime
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.