Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 4-1BBL (CD137L) Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Catalog No. MA558417
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA558417 100 μg
MA558418 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA558417 Supplier Invitrogen™ Supplier No. MA558417
  • $514.00
    $385.65 / Each of 1

    Save $128.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Abbvie patent anti-TNFSF9.

CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-1BBL (CD137L) (Biosimilar)
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Gene TNFSF9
Gene Accession No. P41273
Gene Alias 4 1 BB Ligand; 4 1 BBL; 41BB Ligand; 4-1BB ligand; 41BBL; 4-1BBL; 4-1BB-L; AI848817; CD 137 Ligand; CD 137L; CD137 Ligand; CD137L; Cd157l; CDw137L; costimulatory molecule; homolog of mouse 4-1BB-L; Ly63l; receptor 4-1BB ligand; TNFRSF9; Tnfsf9; TNLG5A; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; tumor necrosis factor ligand superfamily member 9; tumor necrosis factor superfamily member 9
Gene Symbols TNFSF9
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8744
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.