Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4(1H)-Pyrimidinone, 98%

Click to view available options
Quantity:
1g
5g
4562-27-0
C4H4N2O
96.089
MFCD00006664
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
pyrimidin-4-ol,4-hydroxypyrimidine,4-pyrimidinol,4 3h-pyrimidone,pyrimidin-4 3h-one,4 3h-pyrimidinone,4-pyrimidone,4 1h-pyrimidinone,4-pyrimidinone,deaminoisocytosine
20695
1H-pyrimidin-6-one
C1=CN=CNC1=O
4562-27-0
C4H4N2O
96.089
MFCD00006664
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
pyrimidin-4-ol,4-hydroxypyrimidine,4-pyrimidinol,4 3h-pyrimidone,pyrimidin-4 3h-one,4 3h-pyrimidinone,4-pyrimidone,4 1h-pyrimidinone,4-pyrimidinone,deaminoisocytosine
20695
1H-pyrimidin-6-one
C1=CN=CNC1=O
Melting Point 163°C to 169°C (literature)
Quantity 5g
Beilstein 741972
Formula Weight 96.09
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4(1H)-Pyrimidinone

RTECSNumber : UW7350100

TSCA : TSCA

storageNote1 : 2° to 8°C