Learn More
Sigma Organic Chemistry 4-1H-Tetraazol-5-yl ben | 100mg | MFCD01311033
Supplier: Sigma Organic Chemistry CDS001024100MG

4-1H-Tetraazol-5-yl ben , 100mg
About this Item:
MDL #: MFCD01311033
Molecular Weight: 174.16
UNSPSC Code: 12352200
Chemical Formula: C8H6N4O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.