Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2,4-Difluorobenzoyl)piperidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D397925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D397925G 25 g
D39795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D397925G Supplier TCI America Supplier No. D397925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 106266-04-0
Molecular Formula C12H14ClF2NO
Molecular Weight (g/mol) 261.697
MDL Number MFCD01313310
InChI Key QPJONRGTWKXJLG-UHFFFAOYSA-N
Synonym 4-2,4-difluorobenzoyl piperidine hydrochloride, 4-2,4-difluorobenzoyl piperidine hcl, 4-2,4-difluorobenzoyl-piperidine hydrochloride, 2,4-difluorophenyl piperidin-4-yl methanone hydrochloride, 2,4-difluoro-phenyl-piperidin-4-yl-methanone hydrochloride, 2,4-difluorophenyl-4-piperidinyl-methanone hcl, methanone, 2,4-difluorophenyl-4-piperidinyl-, hydrochloride, pubchem16202, acmc-1bvim, 4-2,4-difluorobenzoyl-piperidinehydrochloride
PubChem CID 18759874
IUPAC Name (2,4-difluorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES C1CNCCC1C(=O)C2=C(C=C(C=C2)F)F.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 261.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2,4-Difluorobenzoyl)piperidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.