Learn More
Sigma Organic Chemistry 4- 2 6-Dimethylphenoxy 1G
Supplier: Sigma Organic Chemistry OTV0000961G

4- 2 6-Dimethylphenoxy , 1G
About this item:
MDL #: MFCD06449736
Chemical Formula: C12H16O3
Molecular Weight: 208.25
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.