Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene 95.0+%, TCI America™
SDP

Catalog No. B1565500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1565500G 500 g
B156525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1565500G Supplier TCI America Supplier No. B1565500G

Chemical Identifiers

CAS 5242-49-9
Molecular Formula C29H20N2O2
Molecular Weight (g/mol) 428.491
InChI Key SOTPOQQKAUOHRO-BQYQJAHWSA-N
PubChem CID 5761588
IUPAC Name 2-[4-[(E)-2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-5-methyl-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=CC=CC=C6O5

Specifications

Color Yellow
Quantity 500 g
Formula Weight 428.49
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Benzoxazolyl)-4′-(5-methyl-2-benzoxazolyl)stilbene
EINECSNumber (5)-6068
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.