Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Bromoethyl)benzenesulfonic Acid 97.0+%, TCI America™
SDP

Catalog No. B20061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20061G 1 g
B20065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20061G Supplier TCI America Supplier No. B20061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54322-31-5
Molecular Formula C8H9BrO3S
Molecular Weight (g/mol) 265.121
MDL Number MFCD01861279
InChI Key XRPCDXSRXPQUKT-UHFFFAOYSA-N
PubChem CID 103071
IUPAC Name 4-(2-bromoethyl)benzenesulfonic acid
SMILES C1=CC(=CC=C1CCBr)S(=O)(=O)O

Specifications

Melting Point 86°C
Quantity 1 g
UN Number 2585
Formula Weight 265.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Bromoethyl)benzenesulfonic Acid
EINECSNumber (3)-3831
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.