Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Chloroethyl)morpholine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. C1106500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1106500G 500 g
C110625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1106500G Supplier TCI America Supplier No. C1106500G

Chemical Identifiers

CAS 3647-69-6
Molecular Formula C6H13Cl2NO
Molecular Weight (g/mol) 186.076
MDL Number MFCD00012797
InChI Key NBJHDLKSWUDGJG-UHFFFAOYSA-N
Synonym 4-2-chloroethyl morpholine hydrochloride, n-2-chloroethyl morpholine hydrochloride, 2-morpholinoethylchloride hydrochloride, morpholine, 4-2-chloroethyl-, hydrochloride, 4-2-chloroethyl morpholine hcl, chloromorpholinoethane hydrochloride, 2-chloroethyl morpholine hydrochloride, 2-morpholinoethyl chloride hydrochloride, 4-2-chloroethyl-morpholine hydrochloride, 4-2-chloroethyl morpholinium chloride
PubChem CID 77210
IUPAC Name 4-(2-chloroethyl)morpholine;hydrochloride
SMILES C1COCCN1CCCl.Cl

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 500 g
UN Number 2923
Formula Weight 186.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Chloroethyl)morpholine Hydrochloride
RTECSNumber QE0260000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.