Learn More
Description
- Hydrocinnamaldehyde is c=c double bond hydrogenated cinnamaldehyde. it has been synthesized by the selective hydrogenation of c=c double bond of cinnamaldehyde by various methods. hydrocinnamaldehyde and nitromethane undergoes henry reaction to form nitroaldols. the c1 and c3 position labelled with 13c of hydrocinnamaldehyde was subjected to mass spectrometry and the fragmentation pattern was elucidated.hydrocinnamaldehyde may be used in the synthesis of the following:
- 2-chloro hydrocinnamaldehyde by α-chlorination
- Mixture of homopropargyl alcohols by kinetic resolution-allenylboration reactions
- Mixture of syn- and anti-α-hydroxyallylsilanes by hydroboration of allenylsilane
- (2s)-2-hydroxy-4-phenylbutanenitrile by catalytic asymmetric cyanosilylation
- Cinnamaldehyde by dehydrogenation reaction using pd(tfa)2/4,5-diazafluorenone catalyst
Specifications
Specifications
| CAS | 56705-83-0 |
| Molecular Formula | C13H13NO |
| Linear Formula | C13H13NO |
| MDL Number | MFCD02663364 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 199.25 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.