Learn More
Sigma Organic Chemistry 4-2-Oxiranylmethyl morp | 250mg | 6270-19-5 | MFCD00047415
Supplier: Sigma Organic Chemistry CDS003115250MG

4-2-Oxiranylmethyl morp , 250mg
About this Item:
CAS #: 6270-19-5
MDL #: MFCD00047415
Molecular Weight: 143.18
UNSPSC Code: 12352200
Chemical Formula: C7H13NO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.