Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%

Catalog No. AAH3444106
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3444106 5 g
AAH34441MD 250 mg
AAH3444103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3444106 Supplier Thermo Scientific Chemicals Supplier No. H3444106
Only null left
Add to Cart

CAS: 112539-09-0 | C10H10ClNO3S | 259.704 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112539-09-0
Molecular Formula C10H10ClNO3S
Molecular Weight (g/mol) 259.704
MDL Number MFCD00219329
InChI Key OUAVTZYGHGCCCB-UHFFFAOYSA-N
PubChem CID 2794593
IUPAC Name 4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 104°C to 105°C
Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Synonym 4-2-oxopyrrolidin-1-yl benzenesulfonyl chloride, 4-2-oxo-1-pyrrolidinyl benzenesulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonylchloride, 4-2-oxopyrrolidin-1-yl benzene-1-sulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-oxo-1-pyrrolidinyl, 1-4-chlorosulfonyl phenyl pyrrolidin-2-one, pubchem5680, acmc-1bri4, benzenesulfonylchloride,4-2-oxo-1-pyrrolidinyl
Formula Weight 259.71
Percent Purity 97%
Chemical Name or Material 4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.