Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3,4-Difluorophenyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Catalog No. BTB07014DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB07014DA 1 g Amber Glass Bottle
BTB07014EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB07014DA Supplier Thermo Scientific Chemicals Supplier No. BTB07014DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175135-32-7
Molecular Formula C9H6F2N2S
Molecular Weight (g/mol) 212.218
MDL Number MFCD00052876
InChI Key NDCSJUJQMRFHEX-UHFFFAOYSA-N
Synonym 4-3,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-3,4-difluorophenyl thiazole, 4-3,4-difluorophenyl thiazol-2-amine, 4-3,4-difluoro-phenyl-thiazol-2-ylamine, 2-amino-4-3,4-difluorophenyl-1,3-thiazole, 2-thiazolamine,4-3,4-difluorophenyl, 4-3,4-difluoro-phenyl-1,3-thiazol-2-amine, 4-3,4-difluorophenyl-1,3-thiazole-2-ylamine, pubchem14456, timtec-bb sbb005523
PubChem CID 706326
IUPAC Name 4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
SMILES C1=CC(=C(C=C1C2=CSC(=N2)N)F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 212.22
Percent Purity 97%
Chemical Name or Material 4-(3, 4-Difluorophenyl)-1, 3-thiazol-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.