Learn More
Sigma Organic Chemistry 4- 3 5-Dimethylisoxazol 1G
Supplier: Sigma Organic Chemistry OTV0010011G

4- 3 5-Dimethylisoxazol , 1G
About this item:
MDL #: MFCD02654899
Chemical Formula: C13H13NO4
Molecular Weight: 247.25
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.