Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Chloropropyl)morpholine 98.0+%, TCI America™
SDP

Catalog No. C260225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C260225G 25 g
C26025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C260225G Supplier TCI America Supplier No. C260225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7357-67-7
Molecular Formula C7H14ClNO
Molecular Weight (g/mol) 163.645
MDL Number MFCD00039714
InChI Key PIAZYBLGBSMNLX-UHFFFAOYSA-N
Synonym 4-3-chloropropyl morpholine, n-3-chloropropyl morpholine, morpholine, 4-3-chloropropyl, 3-morpholinopropyl chloride, 1-chloro-3-morpholinopropane, 4-3-chloropropyl-morpholine, 1-chloro-3-morpholin-4-yl propane, 4-3-chloro-propyl-morpholine, morpholine, 4-3-chloropropyl-, hydrochloride
PubChem CID 95834
IUPAC Name 4-(3-chloropropyl)morpholine
SMILES C1COCCN1CCCCl

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 163.65
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(3-Chloropropyl)morpholine
RTECSNumber QE0488000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.