Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Pyridyl)benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. P23321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23321G 1 g
P2332200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23321G Supplier TCI America Supplier No. P23321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127406-55-7
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD01631910
InChI Key NXZUVHZZIZHEOP-UHFFFAOYSA-N
Synonym 4-pyridin-3-yl benzaldehyde, 4-pyridin-3-yl-benzaldehyde, 4-3-pyridyl benzaldehyde, 4-3-pyridinyl benzaldehyde, 4-pyrid-3-ylbenzaldehyde, benzaldehyde, 4-3-pyridinyl, acmc-1byq9, 3-4-formylphenyl pyridine
PubChem CID 3693046
IUPAC Name 4-pyridin-3-ylbenzaldehyde
SMILES C1=CC(=CN=C1)C2=CC=C(C=C2)C=O

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 1 g
Formula Weight 183.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Pyridyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.