Learn More
Sigma Organic Chemistry 4- 3-Trifluoromethyl-be | 1G | MFCD05863563
Supplier: Sigma Organic Chemistry BBO0004451G

4- 3-Trifluoromethyl-be , 1G
About this Item:
MDL #: MFCD05863563
Molecular Weight: 243.27
UNSPSC Code: 12352200
Chemical Formula: C13H16F3N
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.