Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. I0856100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0856100G 100 g
I085625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0856100G Supplier TCI America Supplier No. I0856100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38103-06-9
Molecular Formula C31H20O8
Molecular Weight (g/mol) 520.49
MDL Number MFCD00319167
InChI Key MQAHXEQUBNDFGI-UHFFFAOYSA-N
PubChem CID 94483
IUPAC Name 5-[4-(2-{4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]phenyl}propan-2-yl)phenoxy]-1,3-dihydro-2-benzofuran-1,3-dione
SMILES CC(C)(C1=CC=C(OC2=CC=C3C(=O)OC(=O)C3=C2)C=C1)C1=CC=C(OC2=CC=C3C(=O)OC(=O)C3=C2)C=C1

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 100 g
Synonym 4,4′-(4,4′-Isopropylidenediphenoxy)bis(phthalic Anhydride)
Formula Weight 520.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-(4,4′-Isopropylidenediphenoxy)diphthalic Anhydride
RTECSNumber NP7227500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.