Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole 98.0+%, TCI America™
SDP

Catalog No. T319625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T319625G 25 g
T31965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T319625G Supplier TCI America Supplier No. T319625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 171364-79-7
Molecular Formula C13H19BO3
Molecular Weight (g/mol) 234.10
MDL Number MFCD05663864
InChI Key VFIKPDSQDNROGM-UHFFFAOYSA-N
Synonym 4-Methoxyphenylboronic Acid Pinacol Ester, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 584319
IUPAC Name 2-(4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES COC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.