Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran, Thermo Scientific™

Catalog No. CC29939CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC29939CB 250 mg
CC29939DA 1 g
CC29939DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29939CB Supplier Thermo Scientific Chemicals Supplier No. CC29939CB
Add to Cart

Chemical Identifiers

CAS 1131912-76-9
Molecular Formula C11H21BO3
Molecular Weight (g/mol) 212.10
MDL Number MFCD11506065
InChI Key NLSMOSUUBUCSPL-UHFFFAOYSA-N
Synonym tetrahydropyran-4-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-2h-pyran-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl tetrahydropyran, tetrahydropyran-4-boronic acid, pinacol ester, tetrahydropyran-4-boronicacidpinacolester, 4,4,5,5-tetramethyl-2-oxan-4-yl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl oxane, tetrahydro-2h-pyran-4-yl boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydro-2h-pyran
PubChem CID 42614649
IUPAC Name 4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CCOCC1

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 212.09
Percent Purity 97%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.