Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione 98.0+%, TCI America™
SDP

Catalog No. T299725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T299725G 25 g
T29975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T299725G Supplier TCI America Supplier No. T299725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 720-94-5
Molecular Formula C11H9F3O2
Molecular Weight (g/mol) 230.186
MDL Number MFCD00517909
InChI Key WRZMHTIRFOFFPY-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-p-tolyl butane-1,3-dione, 4,4,4-trifluoro-1-4-methylphenyl butane-1,3-dione, 1-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, l-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, 4,4,4-trifluoro-1-4-methyl phenyl-butane-1,3-dione, 3-4-methylbenzoyl-1,1,1-trifluoroacetone, 4,4,4-trifluoro-1-p-tolylbutane-1,3-dione, 4,4,4-trifluoro-1-p-tolyl-butane-1,3-dione, 1-4-methylphenyl-4,4,4-triflurobutane-1,3-dione, 1,3-butanedione, 4,4,4-trifluoro-1-4-methylphenyl
PubChem CID 550193
IUPAC Name 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione
SMILES CC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F

Specifications

Melting Point 47°C
Quantity 25 g
Formula Weight 230.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.