Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Trihydroxytriphenylmethane, 97%, Thermo Scientific™

Catalog No. AC450100250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450100250 25 g
AC450101000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450100250 Supplier Thermo Scientific Chemicals Supplier No. 450100250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 603-44-1
Molecular Formula C19H16O3
Molecular Weight (g/mol) 292.33
MDL Number MFCD00191589
InChI Key WFCQTAXSWSWIHS-UHFFFAOYSA-N
Synonym 4,4',4-methanetriyltriphenol, leucoaurin, 4,4',4-trihydroxytriphenylmethane, leucoaurine, leucaurin, phenol, methylidynetris, 4-bis 4-hydroxyphenyl methyl phenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, phenol, 4,4',4-methylidynetris
PubChem CID 69047
IUPAC Name 4-[bis(4-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 244.0°C to 246.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 292.33
Percent Purity 97%
Chemical Name or Material 4, 4', 4''-Trihydroxytriphenylmethane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 210-040-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.