Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Tris(benzoyloxy)trityl Bromide 97.0+%, TCI America™
SDP

Catalog No. T107125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T107125G 25 g
T10715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T107125G Supplier TCI America Supplier No. T107125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86610-66-4
Molecular Formula C40H27BrO6
Molecular Weight (g/mol) 683.554
MDL Number MFCD00010302
InChI Key PKXTVZCJOBWKIG-UHFFFAOYSA-N
Synonym Tris[4-(phenylcarbonyloxy)phenyl]methyl Bromide
PubChem CID 3933738
IUPAC Name [4-[bis(4-benzoyloxyphenyl)-bromomethyl]phenyl] benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)C(C3=CC=C(C=C3)OC(=O)C4=CC=CC=C4)(C5=CC=C(C=C5)OC(=O)C6=CC=CC=C6)Br

Specifications

Melting Point 190°C
Quantity 25 g
Formula Weight 683.55
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Tris(benzoyloxy)trityl Bromide [Hydroxyl Protecting Agent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.