Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol 98.0+%, TCI America™
SDP

Catalog No. N1040100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1040100G 100 g
N10405G 5 g
N104025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N1040100G Supplier TCI America Supplier No. N1040100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83310-97-8
Molecular Formula C7H7F9O
Molecular Weight (g/mol) 278.12
MDL Number MFCD02183552
InChI Key OVBNEUIFHDEQHD-UHFFFAOYSA-N
Synonym 3-(Perfluorobutyl)propanol
PubChem CID 2779091
IUPAC Name 4,4,5,5,6,6,7,7,7-nonafluoroheptan-1-ol
SMILES OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 64°C
Quantity 100 g
Formula Weight 278.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.