Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Azo-bis(4-cyanovaleric acid), ∽98%, MP Biomedicals™

Catalog No. ICN15041205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15041205 5 g
ICN15041225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15041205 Supplier MP Biomedicals, Inc Supplier No. 0215041205

Chemical Identifiers

CAS 2638-94-0
Molecular Formula C12H16N4O4
Molecular Weight (g/mol) 280.284
MDL Number MFCD00002799
InChI Key VFXXTYGQYWRHJP-UHFFFAOYSA-N
Synonym 4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
PubChem CID 92938
IUPAC Name 4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
SMILES CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N

Specifications

Color White
Assay Percent Range ∽ 98%
Quantity 5 g
Formula Weight 280.28
Physical Form Powder
Chemical Name or Material 4, 4'-Azo-bis(4-cyanovaleric acid), Polymer synthesis
storageNote1 Protect from light.
Recommended Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.