Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphenyldicarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. B119125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B119125G 25 g
B11915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B119125G Supplier TCI America Supplier No. B119125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 787-70-2
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00002554
InChI Key NEQFBGHQPUXOFH-UHFFFAOYSA-N
Synonym biphenyl-4,4'-dicarboxylic acid, 4,4'-biphenyldicarboxylic acid, 1,1'-biphenyl-4,4'-dicarboxylic acid, 4-4-carboxyphenyl benzoic acid, 4,4'-bibenzoic acid, 4,4'-diphenic acid, 4,4'-dicarboxybiphenyl, p,p'-diphenic acid, usaf do-69, bibenzoic acid
PubChem CID 13084
IUPAC Name 4-(4-carboxyphenyl)benzoic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O

Specifications

Quantity 25 g
Formula Weight 242.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldicarboxylic Acid
RTECSNumber DV2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.