Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(diethylamino)benzophenone 98.0+%, TCI America™
SDP

Catalog No. B0139100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0139100G 100 g
B013925G 25 g
B0139500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0139100G Supplier TCI America Supplier No. B0139100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90-93-7
Molecular Formula C21H28N2O
Molecular Weight (g/mol) 324.47
MDL Number MFCD00009044
InChI Key VYHBFRJRBHMIQZ-UHFFFAOYSA-N
Synonym 4,4'-bis diethylamino benzophenone, michler's ethyl ketone, bis 4-diethylamino phenyl methanone, methanone, bis 4-diethylamino phenyl, 4,4'-tetraethyldiamino benzophenone, p,p'-bis diethylamino benzophenone, p,p'-tetraethyldiamino benzophenone, ethyl michler ketone
PubChem CID 66663
IUPAC Name 4-[4-(diethylamino)benzoyl]-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(CC)CC

Specifications

Melting Point 94°C
Quantity 100 g
UN Number 3077
Formula Weight 324.47
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(diethylamino)benzophenone
EINECSNumber (4)-0893
RTECSNumber OS9545500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.