Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(dodecyloxy)-3-methylazobenzene 93.0+%, TCI America™
SDP

Catalog No. B4598500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4598500MG 500 mg
B45985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4598500MG Supplier TCI America Supplier No. B4598500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1440509-01-2
Molecular Formula C37H60N2O2

Specifications

Quantity 500 mg
Formula Weight 564.90
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(dodecyloxy)-3-methylazobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.