Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Bromophenyl)dibenzothiophene 98.0+%, TCI America™
SDP

Catalog No. B53841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53841G 1 g
B5384200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53841G Supplier TCI America Supplier No. B53841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 530402-77-8
Molecular Formula C18H11BrS
Molecular Weight (g/mol) 339.25
MDL Number MFCD19441307
InChI Key FIAXBPCUAUSGJH-UHFFFAOYSA-N
PubChem CID 59866463
IUPAC Name 4-(4-bromophenyl)dibenzothiophene
SMILES C1=CC=C2C(=C1)C3=C(S2)C(=CC=C3)C4=CC=C(C=C4)Br

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 1 g
Formula Weight 339.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenyl)dibenzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.