Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Bromophenylazo)phenol 98.0+%, TCI America™
SDP

Catalog No. B40531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40531G 1 g
B4053200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40531G Supplier TCI America Supplier No. B40531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3035-94-7
Molecular Formula C12H9BrN2O
Molecular Weight (g/mol) 277.121
MDL Number MFCD00093929
InChI Key BGGPPMHBCFPATP-UHFFFAOYSA-N
Synonym 4-Bromo-4′C-hydroxyazobenzene
PubChem CID 5492867
IUPAC Name 4-[(4-bromophenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES C1=CC(=O)C=CC1=NNC2=CC=C(C=C2)Br

Specifications

Melting Point 161°C
Quantity 1 g
Formula Weight 277.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenylazo)phenol
RTECSNumber SJ8380000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.