Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chloro-2-nitrophenyl)morpholine, 97%, Thermo Scientific™

Catalog No. AAH6305706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6305706 5 g
AAH63057MD 250 mg
AAH6305703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6305706 Supplier Thermo Scientific Chemicals Supplier No. H6305706

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65976-60-5
Molecular Formula C10H11ClN2O3
Molecular Weight (g/mol) 242.66
MDL Number MFCD00455140
InChI Key KJISHWYSNGQNFM-UHFFFAOYSA-N
Synonym 4-4-chloro-2-nitrophenyl morpholine, morpholine, 4-4-chloro-2-nitrophenyl, 1-morpholino-2-nitro-4-chlorobenzene, 4-4-chloro-2-nitro-phenyl-morpholine, 4-4-chloro-2-nitrophenyl morpholine #
PubChem CID 522191
IUPAC Name 4-(4-chloro-2-nitrophenyl)morpholine
SMILES [O-][N+](=O)C1=C(C=CC(Cl)=C1)N1CCOCC1

Specifications

Quantity 5 g
Beilstein 218646
Formula Weight 242.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Chloro-2-nitrophenyl)morpholine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.