Learn More
Sigma Organic Chemistry 4- 4-chlorobenzoyl pipe | 1G | MFCD04113538
Supplier: Sigma Organic Chemistry CDS0032041G

4- 4-chlorobenzoyl pipe , 1G
About this Item:
MDL #: MFCD04113538
Molecular Weight: 223.7
UNSPSC Code: 12352200
Chemical Formula: C12H14ClNO
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.