Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorophenoxy)piperidine,98%

Catalog No. AAH5094403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5094403 1 g
AAH50944MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5094403 Supplier Thermo Scientific Chemicals Supplier No. H5094403
Add to Cart

CAS: 97839-99-1 | C11H14ClNO | 211.69 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97839-99-1
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.69
MDL Number MFCD04113544
InChI Key VOQMPZXAFLPTMM-UHFFFAOYSA-N
PubChem CID 5199889
IUPAC Name 4-(4-chlorophenoxy)piperidine
SMILES ClC1=CC=C(OC2CCNCC2)C=C1

Specifications

Quantity 1 g
Synonym 4-4-chlorophenoxy piperidine, 4-4-chloro-phenoxy-piperidine, piperidine, 4-4-chlorophenoxy, acmc-20amwb, 4-p-chlorophenoxy piperidine, 4-4-chlorophenyl oxy piperidine, 4-chloro-1-4-piperidyloxy benzene, piperidine,4-4-chlorophenoxy, piperidine, 4-4-chlorophenoxy-, hydrochloride
Solubility Information Slightly soluble in water.
Formula Weight 211.69
Percent Purity 98%
Chemical Name or Material 4-(4-Chlorophenoxy)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.