Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-DDE Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USPP1701
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PP-1701 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PP-1701 Supplier Agilent Technologies Supplier No. PP1701
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72-55-9 , 67-56-1
Molecular Formula C14H8Cl4
Molecular Weight (g/mol) 318.02
MDL Number MFCD00000837
InChI Key UCNVFOCBFJOQAL-UHFFFAOYSA-N
PubChem CID 3035
ChEBI CHEBI:16598
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
SMILES ClC(Cl)=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,4,4'-DDE
Element Methanol,4,4'-DDE
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 4, 4'-DDE Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.