Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diamino-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. D28921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28921G 1 g
D2892200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28921G Supplier TCI America Supplier No. D28921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18511-69-8
Molecular Formula C10H10N4
Molecular Weight (g/mol) 186.218
MDL Number MFCD08704188
InChI Key WTHJTVKLMSJXEV-UHFFFAOYSA-N
Synonym 4,4′C-Diamino-2,2′C-bipyridine
PubChem CID 191515
IUPAC Name 2-(4-aminopyridin-2-yl)pyridin-4-amine
SMILES C1=CN=C(C=C1N)C2=NC=CC(=C2)N

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 186.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diamino-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.