Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 4,4'-Diazido-2,2'-stilbenedisulfonic Acid Disodium Salt Tetrahydrate, >99% Purity, Solid, 10 g

Catalog No. 502705252
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5252 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5252 Supplier Abcam Supplier No. AB14537610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Water-soluble reagent for enzyme immobilization on fibroin by photo-crosslinking.

  • Molecular weight: 538.42 g/mol
  • Purity: >99%
  • Form: Solid
  • Key application: Reagent for enzyme immobilisation on fibroin by photo-crosslinking
  • Solubility: Water-soluble over a wide pH range
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Fibroin
Shipping Condition Ambient
Buffer 100% disodium 4,4'-diazidostilbene-2,2'-disulfonate
Form Solid
Content And Storage Store at 4°C.
Formulation 100% Disodium 4,4'-Diazidostilbene-2,2'-disulfonate
Quantity 10 g
For Use With (Application) Enzyme immobilisation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.