Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4″-Dibromo-1,1':2',1″-terphenyl 98.0+%, TCI America™
SDP

Catalog No. D49981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49981G 1 g
D4998200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49981G Supplier TCI America Supplier No. D49981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24253-43-8
Molecular Formula C18H12Br2
Molecular Weight (g/mol) 388.102
MDL Number MFCD28138092
InChI Key CJNMHWJZCGSVQG-UHFFFAOYSA-N
Synonym 4,4′C′C-Dibromo-o-terphenyl
PubChem CID 12715106
IUPAC Name 1,2-bis(4-bromophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br

Specifications

Melting Point 173°C
Quantity 1 g
UN Number 3152
Formula Weight 388.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′′-Dibromo-1,1′:2′,1′′-terphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.