Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol 98.0+%, TCI America™
SDP

Catalog No. D43321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43321G 1 g
D43325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43321G Supplier TCI America Supplier No. D43321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 681481-91-4
Molecular Formula C17H14Br2O2
Molecular Weight (g/mol) 410.105
MDL Number MFCD16660947
InChI Key OZVMVFKUNSNXQC-UHFFFAOYSA-N
PubChem CID 11235237
IUPAC Name 7,7'-dibromo-3,3'-spirobi[1,2-dihydroindene]-4,4'-diol
SMILES C1CC2(CCC3=C(C=CC(=C32)O)Br)C4=C(C=CC(=C41)Br)O

Specifications

Melting Point 148°C
Quantity 1 g
Synonym 4,4′C-Dibromo-7,7′C-dihydroxy-1,1′C-spirobiindane
Formula Weight 410.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-1,1′-spirobiindane-7,7′-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.