Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromo-2,2'-bipyridyl 97.0+%, TCI America™
SDP

Catalog No. D391925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D391925G 25 g
D39191G 1 g
D39195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D391925G Supplier TCI America Supplier No. D391925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18511-71-2
Molecular Formula C10H6Br2N2
Molecular Weight (g/mol) 313.98
MDL Number MFCD00667763
InChI Key KIIHBDSNVJRWFY-UHFFFAOYSA-N
Synonym 4,4'-dibromo-2,2'-bipyridine, 4-bromo-2-4-bromopyridin-2-yl pyridine, 4,4'-dibromo-2,2'-bipyridyl, 2,2'-bipyridine, 4,4'-dibromo, bpbb01, acmc-1bo83, 4,4'-dibromo-2,2-bipyridine, 4,4'-bromo-2,2'-bipyridine, 4,4'-dibromo-2,2'-dipyridine, 4,4'-dibromine-2,2'-bipyridine
PubChem CID 15569987
IUPAC Name 4-bromo-2-(4-bromopyridin-2-yl)pyridine
SMILES C1=CN=C(C=C1Br)C2=NC=CC(=C2)Br

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 25 g
Formula Weight 313.98
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.