Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromo-4″-tert-butyltriphenylamine 95.0+%, TCI America™
SDP

Catalog No. D53301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53301G 1 g
D53305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53301G Supplier TCI America Supplier No. D53301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 852534-22-6
Molecular Formula C22H21Br2N
Molecular Weight (g/mol) 459.225
InChI Key KKLLWAUOZGXWDN-UHFFFAOYSA-N
Synonym N,N-Bis(4-bromophenyl)-4-tert-butylaniline
PubChem CID 20754184
IUPAC Name N,N-bis(4-bromophenyl)-4-tert-butylaniline
SMILES CC(C)(C)C1=CC=C(C=C1)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br

Specifications

Quantity 1 g
Formula Weight 459.23
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Lumps
Chemical Name or Material 4,4′-Dibromo-4′′-tert-butyltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.