Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromobenzil 97.0+%, TCI America™
SDP

Catalog No. D197925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D197925G 25 g
D19795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D197925G Supplier TCI America Supplier No. D197925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35578-47-3
Molecular Formula C14H8Br2O2
Molecular Weight (g/mol) 368.024
MDL Number MFCD00000104
InChI Key NYCBYBDDECLFPE-UHFFFAOYSA-N
Synonym 4,4'-dibromobenzil, ethanedione, bis 4-bromophenyl, p,p'-dibromobenzil, 1,2-bis 4-bromophenyl ethane-1,2-dione, benzyl, 4,4'-dibromo, unii-o4txv8a656, bis 4-bromophenyl ethanedione, 1,2-ethanedione, 1,2-bis 4-bromophenyl, bis 4-bromophenyl ethane-1,2-dione, 1,2-bis-4-bromo-phenyl-ethane-1,2-dione
PubChem CID 96430
IUPAC Name 1,2-bis(4-bromophenyl)ethane-1,2-dione
SMILES C1=CC(=CC=C1C(=O)C(=O)C2=CC=C(C=C2)Br)Br

Specifications

Melting Point 227°C
Quantity 25 g
Formula Weight 368.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromobenzil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.