Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dicarboxydiphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. D2115100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2115100G 100 g
D211525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2115100G Supplier TCI America Supplier No. D2115100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2215-89-6
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.229
MDL Number MFCD00013988
InChI Key WVDRSXGPQWNUBN-UHFFFAOYSA-N
Synonym 4,4'-oxybis benzoic acid, 4,4'-oxydibenzoic acid, 4,4'-oxybisbenzoic acid, 4-4-carboxyphenoxy benzoic acid, benzoic acid, 4,4'-oxybis, 4,4'-dicarboxydiphenyl ether, 4,4-oxobisbenzoic acid, 4,4'-diphenyl ether dicarboxylic acid, diphenyl ether 4,4'-dicarboxylic acid, pubchem9032
PubChem CID 75183
IUPAC Name 4-(4-carboxyphenoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)C(=O)O

Specifications

Color White
Quantity 100 g
Formula Weight 258.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dicarboxydiphenyl Ether
EINECSNumber (3)-4453&(3)-4483
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.