Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichloro-α-methylbenzhydrol 99%, Thermo Scientific™

Catalog No. AC184530100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC184530100 10 g
AC184530500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC184530100 Supplier Thermo Scientific Chemicals Supplier No. 184530100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-06-8
Molecular Formula C14H12Cl2O
Molecular Weight (g/mol) 267.15
MDL Number MFCD00000628
InChI Key URYAFVKLYSEINW-UHFFFAOYSA-N
Synonym chlorfenethol, 4,4'-dichloro-alpha-methylbenzhydrol, dimit, 1,1-bis 4-chlorophenyl ethanol, micasin, mikazene, quickron, qikron, dimite, bcpe
PubChem CID 6624
ChEBI CHEBI:82154
IUPAC Name 1,1-bis(4-chlorophenyl)ethan-1-ol
SMILES CC(O)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Acidity Passes Test
CAS Min % 98.5
CAS Max % 100.0
Melting Point 68°C to 71°C
Color White to Yellow
Infrared Spectrum Authentic
Linear Formula (ClC6H4)2C(CH3)OH
Quantity 10 g
Beilstein 06, III, 3396
Merck Index 12, 2136
Formula Weight 267.14
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Dichloro-α-methylbenzhydrol, 99%
Show More Show Less
EINECSNumber 201-246-3
RTECSNumber DC7875000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.