Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichloro-trans-stilbene 98.0+%, TCI America™
SDP

Catalog No. D30631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30631G 1 g
D3063250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D30631G Supplier TCI America Supplier No. D30631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1657-56-3
Molecular Formula C14H10Cl2
Molecular Weight (g/mol) 249.134
InChI Key WELCIURRBCOJDX-OWOJBTEDSA-N
PubChem CID 5352400
IUPAC Name 1-chloro-4-[(E)-2-(4-chlorophenyl)ethenyl]benzene
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)Cl)Cl

Specifications

Melting Point 176°C
Boiling Point 230°C
Quantity 1 g
Formula Weight 249.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichloro-trans-stilbene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.