Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichlorobenzil 98.0+%, TCI America™
SDP

Catalog No. D44401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44401G 1 g
D44405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44401G Supplier TCI America Supplier No. D44401G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3457-46-3
Molecular Formula C14H8Cl2O2
Molecular Weight (g/mol) 279.116
MDL Number MFCD00018685
InChI Key XMAWUPHYEABFDR-UHFFFAOYSA-N
PubChem CID 18946
IUPAC Name 1,2-bis(4-chlorophenyl)ethane-1,2-dione
SMILES C1=CC(=CC=C1C(=O)C(=O)C2=CC=C(C=C2)Cl)Cl

Specifications

Melting Point 199°C
Color Yellow
Boiling Point 220°C
Quantity 1 g
Synonym 4,4′C-Dichlorobibenzoyl, 4,4′C-Dichlorodibenzoyl
Formula Weight 279.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichlorobenzil
EINECSNumber (4)-0276
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.