Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichlorodiphenyltrichloroethane Solution, 5000μg/mL in Methanol, Ultra Scientific

Catalog No. USEPA1099
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1099 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-EPA-1099 Supplier Agilent Technologies Supplier No. EPA10991
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 50-29-3 , 67-56-1
Molecular Formula C14H9Cl5
Molecular Weight (g/mol) 354.476
InChI Key YVGGHNCTFXOJCH-UHFFFAOYSA-N
PubChem CID 3036
ChEBI CHEBI:16130
IUPAC Name 1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)(Cl)Cl)Cl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material 4, 4'-Dichlorodiphenyltrichloroethane, 5000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.