Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Difluoro-1,3-dimethyl-4-bora-3a,4a-diaza-s-indacene 98.0+%, TCI America™
SDP

Catalog No. D48971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48971G 1 g
D4897200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48971G Supplier TCI America Supplier No. D48971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 154793-49-4
Molecular Formula C11H11BF2N2
Molecular Weight (g/mol) 220.03
InChI Key PIHALWZKFOHBCB-UHFFFAOYSA-N
PubChem CID 124201520
SMILES [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C)(F)F

Specifications

Melting Point 138°C
Quantity 1 g
Formula Weight 220.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Difluoro-1,3-dimethyl-4-bora-3a,4a-diaza-s-indacene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.