Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Difluorobenzophenone 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
345-92-6
C13H8F2O
218.2
MFCD00000353
LSQARZALBDFYQZ-UHFFFAOYSA-N
4,4'-difluorobenzophenone,bis 4-fluorophenyl methanone,bis 4-fluorophenyl-methanone,methanone, bis 4-fluorophenyl,p,p'-difluorobenzophenone,di-p-fluorophenyl ketone,bis p-fluorophenyl ketone,bis 4-fluorophenyl ketone,benzophenone, 4,4'-difluoro,unii-88bnc11b9c
9582
bis(4-fluorophenyl)methanone
C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)F)F
345-92-6
C13H8F2O
218.2
MFCD00000353
LSQARZALBDFYQZ-UHFFFAOYSA-N
4,4'-difluorobenzophenone,bis 4-fluorophenyl methanone,bis 4-fluorophenyl-methanone,methanone, bis 4-fluorophenyl,p,p'-difluorobenzophenone,di-p-fluorophenyl ketone,bis p-fluorophenyl ketone,bis 4-fluorophenyl ketone,benzophenone, 4,4'-difluoro,unii-88bnc11b9c
9582
bis(4-fluorophenyl)methanone
C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)F)F
Boiling Point 200°C
Melting Point 106°C to 109°C
Quantity 100g
Packaging Glass bottle
Loss on Drying 0.25% max.
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (FC6H4)2CO
Formula Weight 218.2
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4, 4'-Difluorobenzophenone, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 206-466-3

RTECSNumber : DJ0685000

TSCA : TSCA