Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihexyl-2,2'-bithiophene 95.0+%, TCI America™
SDP

Catalog No. D41821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41821G 1 g
D4182200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41821G Supplier TCI America Supplier No. D41821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135926-94-2
Molecular Formula C20H30S2
Molecular Weight (g/mol) 334.58
InChI Key RYQPWKFBXWPBGB-UHFFFAOYSA-N
PubChem CID 15540627
IUPAC Name 4-hexyl-2-(4-hexylthiophen-2-yl)thiophene
SMILES CCCCCCC1=CSC(=C1)C2=CC(=CS2)CCCCCC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 334.58
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4′-Dihexyl-2,2′-bithiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.